ContactUsLinkComponent
PKA kinase activity kit
PKA kinase activity kit
  ENZOADIEKS390A
 :  
PKA kinase activity kit
  ENZOADIEKS390A
 :  
  • Pk:
    1 KIT
  • Description:
    PKA kinase activity kit
  • Seq:
    02
  • Size:
    96 wells
The PKA Kinase activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio.
For the quantitative determination of PKA kinase activity in partially purified, purified, or crude enzyme preparations from any species.
 : Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2