Order Entry
Austria
ContactUsLinkComponent
 
Arg8-vasopressin ELISA kit
 
Arg8-vasopressin ELISA kit

 

  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Species independent
  • Target protein:
    Arg8-Vasopressin
  • Sample type:
    Serum, plasma, and tissue culture media.
  • Detection method:
    Colorimetric 450 nm
  • Detection range:
    4,10 - 1000 pg/ml
  • Storage temperature:
    –20/+4 °C
  • Sensitivity:
    2,84 pg/ml

 

Frequently Bought Together