Order Entry
Austria
ContactUsLinkComponent
TSH ELISA kit
TSH ELISA kit
Catalog # ENZOENZKIT1310001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
TSH ELISA kit
Catalog # ENZOENZKIT1310001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    HRP-Streptavidin
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Thyroid Stimulating Hormone (TSH)
  • Description:
    TSH ELISA kit, grow cells
  • Seq:
    01
  • Storage temperature:
    2…8 °C
  • Application:
    Thyroid function
  • Sensitivity:
    0,5 µIU/ml (0,5 - 40 µIU/ml)

Specifications

About this item

Highly sensitive TSH ELISA kit enabling detection of TSH in serum in just 75 minutes.

  • Sensitive measurement of TSH, detecting as little as 0,5 µIU/ml
  • Negligible reactivity from similar glycoprotein hormones
  • High throughput format with results in 75 minutes
  • Fully quantitative results that surpass semi-quantitative Western blot analysis

For the quantitative determination of TSH in serum in human samples.

The TSH ELISA kit is a colorimetric immunometric immunoassay kit with results in 75 minutes.

Ordering information: Contains Microtiter Plate, Standard, Wash Buffer Concentrate, Conjugate, Substrate, and Stop Solution.