Order Entry
Austria
ContactUsLinkComponent
Amp'd® glp-1 elisa kit, grow cells
Amp'd® glp-1 elisa kit, grow cells
Catalog # ENZOENZKIT1040001
Supplier:  Enzo Life Sciences
CAS Number:  
Amp'd® glp-1 elisa kit, grow cells
Catalog # ENZOENZKIT1040001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Target protein:
    GLP-1
  • Description:
    AMP'D® GLP-1 ELISA kit, grow cells
  • Seq:
    01
  • Sensitivity:
    5,5 pg/ml (range 7,8 - 250 pg/ml)

Specifications

About this item

Ultra-sensitive AMP'D® GLP-1 ELISA kit enabling the ability to use less sample to detect levels of human Glucagon-like peptide 1 (7-36) amide, a potent promoter of insulin secretion, a major incretin hormone, and therapeutic for type 2 diabetes.

  • Ultra-sensitive measurement of GLP-1, detecting as little as 5,5 pg/ml
  • Negligible reactivity from similar GLP-1 forms
  • High throughput format with results in three hours for up to 40 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis

For the quantitative determination of Glucagon-like peptide 1 (GLP-1) (7-36) amide in serum and plasma of human origin.

The AMP'D® GLP-1 ELISA kit is a colorimetric immunometric enzyme immunoassay kit with results in three hours.

Ordering information: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Conjugate, and AMP'D® (Signal Amplification) Substrate, Amplifier, Substrate Diluent, Amplifier Diluent, and Stop Solution.