Order Entry
Austria
ContactUsLinkComponent
Amp'd® HSP70 high sensitivity ELISA Kit
Amp'd® HSP70 high sensitivity ELISA Kit
Catalog # ENZOENZKIT1010001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Amp'd® HSP70 high sensitivity ELISA Kit
Catalog # ENZOENZKIT1010001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Host:
  • Primary antibody reactivity:
  • Target protein:
    HSP70
  • Description:
    Amp'd® HSP70 high sensitivity ELISA kit
  • Seq:
    01
  • Environmentally Preferable:
  • Sensitivity:
    7 pg/ml (range 39 - 500 pg/ml)

Specifications

About this item

Ultra-sensitive (7 pg/ml)Amp'd® HSP70 high sensitivity ELISA kit enabling the ability to use less sample and detect both baseline and upregulated levels of human, mouse and rat Hsp70 (Hsp72), a major chaperone, cancer biomarker, and key cell stress regulator.

For the quantitative determination of normal and upregulated (stressed) levels of HSP70 in serum and plasma of human, mouse, and rat origin.

The Amp'd® HSP70 high sensitivity ELISA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4,5 hours.

Ordering information: Contains Microtiter Plate, Assay Buffer 28, Standard, Wash Buffer Concentrate, Antibody, Conjugate, and Amp'd (Signal Amplification) Substrate, Amplifier, Substrate Diluent, Amplifier Diluent, and Stop Solution.