Order Entry
Austria
ContactUsLinkComponent
Amp'd® ELISA signal amplification kit
Amp'd® ELISA signal amplification kit
Supplier:  ENZO LIFE SCIENCES
Certificates
Amp'd® ELISA signal amplification kit
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Conjugate ELISA:
    Alkaline Phosphatase (AP)
  • Detection method:
    Signal amplification 495 nm
  • Time to Results:
    30 minutes
  • Sensitivity:
    Improves ELISA sensitivity ~10 to 50-fold

Specifications

Frequently Bought Together