IgG2b (mouse), ELISA kit
Catalog # ENZOADI900110
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Mouse
- Target protein:IgG2b
- Description:IgG2b (mouse), ELISA kit
- Seq:02
- Sensitivity:1,05 ng/ml (range 7,81 - 250 ng/ml)
Specifications
About this item
For the quantitative determination of IgG2b in culture supernatants and serum of mouse origin.
The IgG2b (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 90 minutes. Ideal for determining the isotype and concentration of monoclonal antibodies.
Ordering information: Contains GxM IgG Microtiter plate, mouse IgG2b conjugate, Assay buffer 13 concentrate, Wash buffer concentrate, mouse IgG2b standard, TMB Substrate, Stop solution 2.