TNF alpha (mouse) ELISA kit
Catalog # ENZOADI900047
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Mouse
- Target protein:TNF-alpha
- Description:TNF-α (mouse), ELISA kit
- Seq:02
- Sensitivity:3,9 pg/ml (range 31,25 - 2000 pg/ml)
Specifications
About this item
For the quantitative determination of mouse TNF-α in culture supernatants and serum. Cited sample type includes tissue and plasma.
The TNF-α (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 5 hours. Absorbance is read at 450 nm.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 16, Wash buffer concentrate, Standard, TMB Substrate, Stop solution.