Order Entry
Austria
ContactUsLinkComponent
 
Progesterone ELISA Kit
  ENZOADI900011
 :  
undefined
Progesterone ELISA Kit
  ENZOADI900011
 :  

 

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Description:
    Progesteron, ELISA kit, 96 wells
  • Seq:
    02
  • Sensitivity:
    8,57pg/ml (range 15,62 - 500 pg/ml)

 

 

For the quantitative determination of progesterone in culture supernatants, serum, and saliva from any species. Cited sample type includes plasma.

The Progesterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in <3 hours. Absorbance is read at 405 nm. This non-radioactive enzyme immunoassay is safe and reliable with ready-to-use liquid color-coded reagents to reduce errors.

 : Contains GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent. Alternative size available: 89141-280 (5x96 wells).