DHEA ELISA kit
Catalog # ENZOADI901093
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:1 KIT
- Assay Type:Competitive
- Host:
- Primary antibody reactivity:
- Description:DHEA ELISA kit
- Seq:02
- Environmentally Preferable:
- Sensitivity:2,9 pg/ml (range 12,21 - 50,000 pg/ml)
Specifications
About this item
For the quantitative determination of DHEA in culture supernatants, plasma, serum, saliva, and urine from any species.
The DHEA EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. This kit has low reactivity to structurally related molecules and offers sensitive measurement of DHEA in a variety of sample types.
Ordering information: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-142 (1x96 wells).