Order Entry
Austria
ContactUsLinkComponent
11-dehydro-TXB2 ELISA Kits
11-dehydro-TXB2 ELISA Kits
Catalog # ENZOADI901092
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
11-dehydro-TXB2 ELISA Kits
Catalog # ENZOADI901092
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    11-dehydro-TXB2
  • Description:
    11-dehydro-TXB2 ELISA kit
  • Seq:
    02
  • Sensitivity:
    4,31 pg/ml (range 9,8 - 10,000 pg/ml)

Specifications

About this item

For the quantitative determination of 11-dehydro-TXB2 in culture supernatants and urine from any species. Cited samples include serum.

The 11-dehydro-TXB2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit offers ultrasensitive quantification of 11-d-TXB2 in a variety of sample matrices with negligible reactivity with related molecules in a convenient 96-well format offers higher throughput analysis compared to conventional analytical methods.

Ordering information: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-140 (1x96 wells).