15(S)-HETE ELISA kit
ENZOADI901051
:
- Pk:1 KIT
- Assay Type:Competitive
- Target protein:15(S)-HETE
- Description:15(S)-HETE ELISA kit
- Seq:02
- Sensitivity:69,21 pg/ml (range 78,1 - 20,000 pg/ml)
For the quantitative determination of 15(S)-HETE in culture supernatants, plasma, serum, and urine from any species.
The 15(S)-HETE EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour.
: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-104 (1x96 wells).