Progesterone ELISA Kit
ENZOADI901011
:
- Pk:1 KIT
- Assay Type:Competitive
- Description:Progesterone ELISA kit
- Seq:02
- Sensitivity:8.57 pg/ml (range 15,62 - 500 pg/ml)
For the quantitative determination of progesterone in culture supernatants, serum, and saliva from any species. Cited sample type includes plasma.
The Progesterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. This non-radioactive enzyme immunoassay is safe and reliable with ready-to-use liquid color-coded reagents to reduce errors.
: Contains GxM Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent. Alternative size available: 89141-064 (1×96 wells).