Order Entry
Austria
ContactUsLinkComponent
PGE2 ELISA kits
PGE2 ELISA kits
Supplier:  ENZO LIFE SCIENCES
Certificates
undefined
PGE2 ELISA kits
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Assay Type:
    Competitive
  • Conjugate ELISA:
    Alkaline Phosphatase (AP)
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Species independent
  • Target protein:
    PGE2
  • Detection method:
    Colorimetric detection 405 nm
  • Time to Results:
    <3 hrs
  • Sensitivity:
    13,4 pg/ml (range 39,1 - 2,500 pg/ml)

Specifications

Frequently Bought Together