Order Entry
Austria
ContactUsLinkComponent
Cu/Zn-Superoxide dismutase ELISA kit
Cu/Zn-Superoxide dismutase ELISA kit
Catalog # ENZOALX850033KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Cu/Zn-Superoxide dismutase ELISA kit
Catalog # ENZOALX850033KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Description:
    Cu/Zn-Superoxide Dismutase ELISA kit
  • Seq:
    02
  • Sensitivity:
    0,04 µg/ml (Range 0,08 to 5 µg/ml)
  • Immunogen:
    Cu/ZnSOD

Specifications

About this item

For the quantitative determination of human Cu/Zn SOD in cell culture supernatants, serum and other body fluids.

  • Assay Time: <2 hours

Late stages of apoptosis are characterized by the loss of membrane integrity and a release of the intracellular Cu/Zn SOD.