COX-2 (human), ELISA kit
Catalog # ENZOADI900094
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:COX-2
- Description:COX-2 (Human), ELISA kit
- Seq:02
- Storage conditions:4 °C (long term)
- Sensitivity:0,25 ng/ml (range 1,09 - 70 ng/ml)
Specifications
About this item
The COX-2 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 2 hours. This is the most sensitive COX-II immunoassay kit commercially available for inflammation research.
- Easy to use
- lThe color-coded reagents reduce errors
- Reliable and fast
For the quantitative determination of human COX-2 in cell lysates.
Delivery: Supplied with microtiter plate, standard, antibody, conjugate, assay buffer 28, TMB substrate, stop solution 2, wash buffer concentrate.