Order Entry
Austria
ContactUsLinkComponent
 
LEADING LIGHT® Wnt cell line assay reagents
Catalog # ENZOENZ600040001
Supplier:  Enzo Life Sciences
CAS Number:  
LEADING LIGHT® Wnt cell line assay reagents
Catalog # ENZOENZ600040001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    LEADING LIGHT® Wnt cell line assay reagents
  • Seq:
    02
  • Size:
    1 Pack

Specifications

About this item

For use with the LEADING LIGHT® Wnt Reporter Cell Line.

  • Can be used as a supplement to the Wnt LEADING LIGHT® Wnt Reporter Assay Starter Kit (Prod. No. ENZ-61001)
  • When used in the Wnt LEADING LIGHT® Wnt Reporter Assay Starter Kit, there are many advantages of using this line of products:
  • True end-point detection system
  • High sensitivity (Wnt3a EC50 = 45.9 ng/ml)
  • Excellent reproducibility (Z'-factor of 0.74)
  • High signal-to-noise ratio without the need for Lithium Chloride to boost the signal
  • High-throughput screening applications (including 384-well microplates)

The LEADING LIGHT® Wnt Cell Line Assay Reagents are intended for use with the LEADING LIGHT® Wnt Reporter Cell Line, which is the basis for a cell-based luciferase activity test ENZ-61001 LEADING LIGHT® Wnt Reporter Assay Starter Kit suitable for a 96-well plate format. The system contains an engineered 3T3 mouse fibroblast cell line, which expresses the firefly luciferase reporter gene under the control of Wnt-responsive promoters (TCF/LEF). The luciferase activity from the reporter gene in this cell line can be up-regulated in a dose-dependent manner upon the addition of exogenous Wnt protein/Wnt agonist or down-regulated by a further addition of a Wnt antagonist to the cell culture medium. This system can be used to elucidate the functions/activities of different Wnt-related ligands such as Wnt, DKK, etc. This system can also be used for screening small molecules and antibodies for their ability to act as Wnt inhibitors or Wnt agonists. The assay has been used successfully in different assay formats (including HTS applications) to identify several distinct categories of small molecule compounds that modulate the Wnt signaling pathway.

The LEADING LIGHT® Wnt Cell Line Assay Reagents contains Luciferase substrate and assay buffer, Lithium Chloride, HEPES, Wnt3a protein (positive control), Dkk-1 protein, and a 96-well black plate.

Note: The complete LEADING LIGHT® Wnt Reporter Assay Starter Kit (Prod. No. ENZ-61001) is available.