Order Entry
Austria
ContactUsLinkComponent
Nitric Oxide (NO2-/NO3-) detection kit
Nitric Oxide (NO2-/NO3-) detection kit
Catalog # ENZOADI917010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Nitric Oxide (NO2-/NO3-) detection kit
Catalog # ENZOADI917010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Nitric Oxide (NO2-/NO3-) detection kit
  • Seq:
    02
  • Sensitivity:
    (Nitrite) 0,222 µmole/L; (Nitrate) 0,625 µmole/L (range (Nitrite) 3,125 - 200 µmole/L; (Nitrate) 3,125 - 100 µmole/L)

Specifications

About this item

For the quantitative determination of nitrite and nitrate levels in culture supernatants, plasma, serum, saliva, and urine from any species.

The Nitric Oxide (NO2-/NO3-), detection kit is a complete kit for the quantitative determination of nitrate and nitrite concentrations. Absorbance is read at 540-570 nm. Only 0.050 mL diluted sample needed per well saves precious sample.

Ordering information: Contains Microtiter plate, Nitrate Reductase, Nitrate Reductase Storage Buffer, NADH, Nitrate Standard, Nitrite Standard, Reaction buffer concentrate, Griess Reagent I, Griess Reagent II.