Order Entry
Austria
Orders LinkContactUsLinkComponent
sPLA2 activity kit

sPLA2 activity kit

sPLA₂ activity kit, 1 KIT
Catalog # ENZOADI907002
Supplier:   Enzo Life Sciences
sPLA2 activity kit
Catalog # ENZOADI907002
Supplier:  Enzo Life Sciences
CAS Number:  
sPLA2 activity kit

sPLA₂ activity kit, 1 KIT

CAS Number:  
Price
634,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

For the quantitative determination of sPLA2 in culture supernatants, cell lysates, and buffers from any species.

  • Higher throughput - assay up to 42 samples in duplicate per 96 well kit
  • Sensitive - detect < 4 U/ml
  • Fast - results in just 40 minutes

The sPLA2 activity kit is a higher throughput assay for the quantification of up to 42 samples in duplicate per 96 well kit. This kit provides safe, sensitive, and reliable results in under an hour.

Ordering information: Contains Microtiter plate, Standard, Reaction buffer concentrate, Substrate, Stop reagent, Color reagent.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    sPLA₂ activity kit
  • Cat. No.:
    ENZOADI907002
  • Sensitivity:
    3,85 Units/ml (range 5 - 80 Units/ml)

Specifications