Order Entry
Austria
ContactUsLinkComponent
LYSO-ID® red cytotoxicity kit (GFP-CERTIFIED®) for microplates
LYSO-ID® red cytotoxicity kit (GFP-CERTIFIED®) for microplates
Catalog # ENZOENZ51015KP002
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
LYSO-ID® red cytotoxicity kit (GFP-CERTIFIED®) for microplates
Catalog # ENZOENZ51015KP002
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    LYSO-ID® Red cytotoxicity kit (GFP-CERTIFIED®)
  • Seq:
    02
  • Tests per kit:
    2× 96-well plates

Specifications

About this item

A Rapid, Quantitative and HTS‐compatible Live Cell Cytotoxicity Assay

  • Assay includes unique drug-like dye that rapidly partitions into cells and labels acidic organelles
  • Only commercial assay available that allows for long-term cell monitoring of cytotoxic effects
  • Multi-well, high-throughput with rapid 10-15 minute dye incubation
  • No co-incubation with artificial phospholipid analogs required for detection, eliminating the potential for confounding dye-associated artifacts
  • Monitors lysosome accumulation as a response to prolonged drug treatment
  • Quantitative results in as little as 3 hours, including drug treatment

96-well cell-based assay. Provides a rapid and quantitative approach for determining drug- or toxic agent-induced lysosome and lysosome-like organelle perturbations in live cells.

Ordering information: Quantity: 2×96-well plates Contents: 10x dual color detection reagent: 2×1 ml
Detection buffer: 20 ml
Verapamil control: 3 µmoles
10x assay buffer: 15 ml