Order Entry
Austria
ContactUsLinkComponent
Myeloperoxidase fluorometric detection kit
Myeloperoxidase fluorometric detection kit
Catalog # ENZOADI907029
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Myeloperoxidase fluorometric detection kit
Catalog # ENZOADI907029
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    500 Tests
  • Description:
    Myeloperoxidase fluorometric detection kit
  • Seq:
    02
  • Storage conditions:
    4 °C (long term)
  • Sensitivity:
    0,00195 units/ml
  • Size:
    500 tests

Specifications

About this item

The Myeloperoxidase fluorometric detection kit is a one-step no wash homogeneous activity assay that is adaptable to HTS applications. This kit offers highly sensitive fluorescent detection and the ability to monitor multiple time points to follow the kinetics of MPO activity.

  • Fast homogenous assay
  • Suitable for High Throughput Screening
  • Monitor multiple time points to follow kinetics
  • Results in 30 to 60 minutes

For the measurement of Myeloperoxidase activity in cells and tissue from any species.

Delivery: Assay buffer concentrate, Detection Reagent, Hydrogen peroxide, Myeloperoxidase enzyme