Order Entry
Austria
ContactUsLinkComponent
Anti-IL7R Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: R34-34]
Anti-IL7R Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: R34-34]
Catalog # ENZOENZABS2510100
Supplier:  ENZO LIFE SCIENCES
Anti-IL7R Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: R34-34]
Catalog # ENZOENZABS2510100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    IL-7 receptor-alpha
  • Antigen symbol:
    IL7R
  • Clonality:
    Monoclonal
  • Clone:
    R34-34
  • Conjugation:
    Alexa Fluor® 647
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Seq:
    02
  • Form:
    Liquid
  • Antigen synonyms:
    IL-7RA|Interleukin-7 receptor α|CD127
  • Storage buffer:
    In PBS containing 50% glycerol.
  • Storage temperature:
    –80 °C
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Human leukemic pre-B cells.
  • Pk:
    100 µG

Specifications

About this item

Recommended Applications: Flow Cytometry

Species reactivity: Human, Dog

Type: Primary
Antigen: IL7R
Clonality: Monoclonal
Clone: R34-34
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human