Anti-FKBP59 Mouse Monoclonal Antibody [clone: KN382/EC1]
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
FKBP59, also known as Hsp56/p59 is an immunophilin that is a component of Hsp70/Hsp90/steroid receptor complexes. Expression of FKBP59 is increased in response to heat shock and ischemic stress, and is thought to play a role in the folding and cytoplasmic/nuclear distribution of steroid receptors.
Type: Primary
Antigen: FKBP59
Clonality: Monoclonal
Clone: KN382/EC1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Monkey, Bovine, Hamster, Mink, Rabbit, Guinea Pig
Specifications
- Catalog No:
- ENZOADISRA1400F
- ENZOADISRA1400D
- Antigen symbol:
- FKBP59
- FKBP59
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Form:
- -
- Liquid. In PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide.
- Purification:
- -
- Protein G affinity purified
Specifications
Frequently Bought Together