Order Entry
Austria
ContactUsLinkComponent
 
Anti-KDEL Mouse Monoclonal Antibody [clone: 10C3]
 
undefined
Anti-KDEL Mouse Monoclonal Antibody [clone: 10C3]

 

  • Catalog No:
  • ENZOADISPA827J
  • ENZOADISPA827F
  • ENZOADISPA827D
  • Antigen symbol:
  • KDEL
  • KDEL
  • KDEL
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • 10C3
  • 10C3
  • 10C3
  • Reactivity:
  • Mammals
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG2a
  • IgG2a
  • IgG2a
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Liquid
  • Purification:
  • Protein G affinity purified
  • Protein G affinity purified
  • Protein G affinity purified
  • Storage buffer:
  • PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide

 

Frequently Bought Together