Anti-HSP90 Mouse Monoclonal Antibody [clone: 2D11B9]
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Type: Primary
Antigen: HSP90
Clonality: Monoclonal
Clone: 2D11B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat, Sheep, Pig, Monkey, Horse, Hamster, Guinea pig, Fish, Drosophila, Dog
Specifications
- Catalog No:
- ENZOADISPA831200
- ENZOADISPA831050
- Antigen symbol:
- HSP90
- HSP90
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 2D11B9
- 2D11B9
- Reactivity:
- Fish, Mouse, Hamster, Drosophila, Pig, Horse, Monkey, Sheep, Human, Guinea pig, Dog, Rat
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- Immunogen:
- Recombinant human Hsp90
- Recombinant human Hsp90
- Form:
- Liquid
- Liquid
- Purification:
- Purified by Protein G affinity chromatography
- Purified by Protein G affinity chromatography
- Storage buffer:
- PBS containing 50% glycerol and 0,09% sodium azide
- PBS containing 50% glycerol and 0,09% sodium azide
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together