Specifications
- Antibody type:Primary
- Antigen symbol:DJ-1
- Clonality:Monoclonal
- Clone:3E8
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Form:Liquid
- Storage buffer:PBS, pH 7,2, containing 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human DJ-1
- Purification:Purified by Protein A affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Researchers cloned DJ-1 protein, consisting of 189 amino acids, from a HeLa cell cDNA library as a oncogene which weakly transformed NIH3T3 cells. Cotransfection with ras or c-myc enhances transformation by DJ-11. DJ-1 expression is induced by growth signals, and it is translocated from the cytoplasm to the nucleus during the S phase without the presence of any nuclear localization signals. In addition to its potential involvement in ras-mediated signal transduction pathways, DJ-1 is known to regulate transcription of androgen-responsive genes, and is sensitive to oxidative stress. DJ-1 is ubiquitously expressed in various human tissues including the brain, with high expression in astrocytes of the frontal cortex and substantia nigra. Homozygous mutations in the DJ-1 gene have been shown to cause early-onset autosomal recessive Parkinson’s disease.
Type: Primary
Antigen: DJ-1
Clonality: Monoclonal
Clone: 3E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human