Order Entry
Austria
ContactUsLinkComponent
 
Anti-PI3 Kinase Mouse Monoclonal Antibody [clone: AB6]
  ENZOADIKAMPI200E
Anti-PI3 Kinase Mouse Monoclonal Antibody [clone: AB6]
  ENZOADIKAMPI200E

 

  • Antibody type:
    Primary
  • Antigen name:
    PI3 Kinase
  • Clonality:
    Monoclonal
  • Clone:
    AB6
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid
  • Antigen synonyms:
    p85 alpha
  • Storage buffer:
    PBS, pH 7,2, containing 50% glycerol
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant human PI3 kinase (p85alpha)
  • Purification:
    Purified by Protein A affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Phosphatidylinositol 3-kinase (PI3-K) is a lipid kinase that phosphorylates the inositol ring of phosphatidylinositol and related compounds at the 3

Type: Primary
Antigen: PI3 kinase
Clonality: Monoclonal
Clone: AB6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat