Order Entry
Austria
ContactUsLinkComponent
Anti-ACTA2 Mouse Monoclonal Antibody
Anti-ACTA2 Mouse Monoclonal Antibody
  ENZOENZC34933
Anti-ACTA2 Mouse Monoclonal Antibody
  ENZOENZC34933

 

  • Antibody type:
    Primary
  • Antigen name:
    Smooth Muscle Actin
  • Antigen symbol:
    ACTA2
  • Clonality:
    Monoclonal
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 0,1% sodium azide and 1% BSA.
  • Antigen synonyms:
    AAT6|alpha 2 actin|MYMY5|aortic smooth muscle|cell growth-inhibiting gene 46 protein|alpha-cardiac actin|alpha-actin-2|actin|GIG46|growth-inhibiting gene 46|ACTSA
  • Purification:
    Affinity purified
  • Pk:
    0,5 mL

 

 

Host: Mouse, Isotype: IgG2b

  • For Immunohistochemistry, Western Blot

Recognizes human, monkey, chicken and rabbit actin isotypes alpha and gamma of smooth muscle cells. Does not cross-react with actin of skeletal or cardiac muscle cells.

Type: Primary
Antigen: ACTA2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human