Anti-Insulin Receptor Rabbit Polyclonal Antibody
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Host: Rabbit
- For Western Blot
Recognizes human and mouse insulin receptor/IGF-1R phosphorylated at Tyr1158/1162/1163.
Type: Primary
Antigen: INSULIN RECEPTOR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- ENZOBMLSA3920100
- ENZOBMLSA3920020
- Antigen symbol:
- INSR
- INSR
- Antigen name:
- Insulin Receptor
- Insulin Receptor
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Form:
- Liquid. In PBS, pH 7,4, containing BSA and sodium azide.
- Liquid. In PBS, pH 7,4, containing BSA and sodium azide.
Specifications
Frequently Bought Together