Anti-BAK Mouse Monoclonal Antibody
ENZOBMLSA2980050
- Antibody type:Primary
- Antigen name:BCL2-Antagonist/Killer 1
- Antigen symbol:BAK
- Clonality:Monoclonal
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Western blot:Yes
- Seq:02
- Form:Liquid, in PBS containing sodium azide and gelatin
- Antigen synonyms:bcl-2 homologous antagonist/killer|bcl-2-like protein 7|BCL2-like 7 protein|pro-apoptotic protein BAK|BAK1|bcl2-L-7|apoptosis regulator BAK
- Purification:Purified
- Pk:50 µG
Host: Mouse, Isotype: IgG2a
- For Immunocytochemistry, Western Blot
Recognizes human Bak.
Type: Primary
Antigen: BAK
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human