Order Entry
Austria
ContactUsLinkComponent
 
Anti-MAPK Rabbit Polyclonal Antibody
  ENZOBMLSA2660020
Anti-MAPK Rabbit Polyclonal Antibody
  ENZOBMLSA2660020

 

  • Antibody type:
    Primary
  • Antigen name:
    p38 MAPK
  • Antigen symbol:
    MAPK
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid, in PBS containing BSA and sodium azide
  • Antigen synonyms:
    MAX-interacting protein 2|MAP kinase MXI2|CSAID-binding protein|MAPK 14|Stress-activated protein kin|CSBP|MAP kinase p38 alpha|Mitogen-activated protein kinase 14|MAP kinase 14|Mitogen-activated protein kinase p38 alpha|Cytokine suppressive anti-inflammatory drug-binding protein
  • Pk:
    20 µl

 

 

Host: Rabbit

  • For Western Blot

Recognizes human, mouse, rat, dog and carp p38 isoforms p38alpha, p38beta, and p38gamma phosphorylated dually at Thr180 and Tyr182. Does not cross-react with unphosphorylated forms of p38.

Type: Primary
Antigen: MAPK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human