Order Entry
Austria
ContactUsLinkComponent
Anti-Y Mouse Monoclonal Antibody
Anti-Y Mouse Monoclonal Antibody
  ENZOBMLSA2401000
Anti-Y Mouse Monoclonal Antibody
  ENZOBMLSA2401000

 

  • Antibody type:
    Primary
  • Antigen name:
    Tyrosine
  • Antigen symbol:
    Y
  • Clonality:
    Monoclonal
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Western blot:
    Yes
  • Seq:
    02
  • Antigen synonyms:
    p-Tyr|Phosphotyrosine|tyr|Phospho tyr|p-tyrosine
  • Purification:
    Purified
  • Pk:
    1 mg

 

 

Host: Mouse, Isotype: IgG2b

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes phosphotyrosine. Species independent. Does not cross-react with phosphoserine or phosphothreonine.

Type: Primary
Antigen: Y
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Many Species