Anti-USP21 Rabbit Polyclonal Antibody
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
USP21 is a ubiquitin-specific protease that catalyzes the hydrolysis of ubH2A thereby acting to relieve ubH2A-specific repression. USP21 is also thought to be involved with hepatocyte regeneration. Furthermore, in addition to its deubiquitinylating activity, USP21 is capable of removing NEDD8 from NEDD8 conjugates but has no effect on SUMO conjugates.
- For Western Blot
Recognizes human USP21.
Type: Primary
Antigen: USP21
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- ENZOBMLPW05850025
- ENZOBMLPW05850100
- Antigen symbol:
- USP21
- USP21
- Antigen name:
- Ubiquitin specific peptidase 21
- Ubiquitin specific peptidase 21
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Human
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Form:
- Liquid, in PBS containing 0,01% sodium azide
- Liquid, in PBS containing 0,01% sodium azide
- Purification:
- Salt precipitation
- Salt precipitation
Specifications
Frequently Bought Together