Anti-HSF1 Rabbit Polyclonal Antibody
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
HSFs (Heat Shock family of transcription factors), which consists of HSF 1-4, bind to highly conserved Heat shock elements (HSEs) in the promoter regions of heat shock genes, ultimately regulating the expression of Heat shock proteins (Hsps). On exposure to heat shock and other stresses, HSF1 localizes within seconds to discrete nuclear granules and on recovery from stress, HSF1 rapidly dissipates from the stress granules to a diffuse nucleoplasmic distribution.
- For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, EMSA
Recognizes human, mouse, rat, gerbil, monkey and rabbit HSF1. Detects a band of ~95kDa by Western blot.
Type: Primary
Antigen: HSF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- ENZOADISPA901J
- ENZOADISPA901F
- ENZOADISPA901D
- Antigen symbol:
- HSF1
- HSF1
- HSF1
- Antigen name:
- Heat Shock Transcription Factor 1
- Heat Shock Transcription Factor 1
- Heat Shock Transcription Factor 1
- Antigen synonyms:
- HSTF 1|Heat shock transcription factor 1|Heat shock factor protein 1|HSTF1|HSF 1
- HSTF 1|Heat shock transcription factor 1|Heat shock factor protein 1|HSTF1|HSF 1
- HSTF 1|Heat shock transcription factor 1|Heat shock factor protein 1|HSTF1|HSF 1
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Reactivity:
- Human
- Host:
- Rabbit
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Yes
- Form:
- Liquid, in PBS containing 50% glycerol and 0,09% sodium azide
- Liquid, in PBS containing 50% glycerol and 0,09% sodium azide
- Liquid, in PBS containing 50% glycerol and 0,09% sodium azide
- Purification:
- Protein A affinity purified
- Protein A affinity purified
- Protein A affinity purified
Specifications
Frequently Bought Together