Order Entry
Austria
ContactUsLinkComponent
 
Anti-HMOX2 Rabbit Polyclonal Antibody
 
Anti-HMOX2 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • ENZOADISPA894F
  • ENZOADISPA894D
  • Antigen symbol:
  • HMOX2
  • HMOX1
  • Antigen name:
  • Heme Oxygenase 2
  • Heme Oxygenase 1
  • Antigen synonyms:
  • Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Reactivity:
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid, in PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • Liquid, in PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • Purification:
  • Protein A affinity purified
  • Protein A affinity purified

 

Frequently Bought Together