Anti-PDI Rabbit Polyclonal Antibody
ENZOADISPA890D
- Antibody type:Primary
- Antigen name:Protein Disulphide-Isomerase
- Antigen symbol:PDI
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Seq:02
- Form:Liquid, in PBS containing 50% glycerol and 0,09% sodium azide
- Antigen synonyms:protein disulfide isomerase|Eps1p|PDI1|CaO19.12595|PDI1P
- Purification:Protein A affinity purified
- Pk:50 µG
The mammalian PDI (Protein disulfide-isomerase) family encompasses several highly divergent proteins which are involved in the processing and maturation of secretory proteins in the endoplasmic reticulum by catalyzing the rearrangement of disulfide bonds.
- For Immunocytochemistry, Immunoprecipitation, Western Blot
Recognizes human, mouse, rat, bovine, dog, guinea pig, hamster, monkey, pig, sheep and Xenopus PDI. Detects a band of ~58kDa by Western blot.
Type: Primary
Antigen: PDI
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human