About this item
Calnexin (CNX), an unglycosylated resident ER transmembrane protein, together with Calreticulin (CRT), plays a key role in glycoprotein folding and its control within the ER, by interacting with folding intermediates via their monoglucosylated glycans. Calnexin associates with newly synthesized monomeric glycoproteins and only recognizes glycoproteins when they are incompletely folded. Furthermore, Calnexin has been demonstrated to function as a molecular chaperone capable of interacting with polypeptide segments of folding glycoproteins.
- For Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot, EM
Recognizes human, mouse, rat, bovine, dog, chicken, Drosophila, guinea pig, hamster, monkey, pig, quail, rabbit, sheep and Xenopus Calnexin. Detects a band of ~90kDa by Western blot.
Type: Primary
Antigen: CANX
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- ENZOADISPA860F
- ENZOADISPA860D
- Antigen symbol:
- CANX
- CANX
- Antigen name:
- Calnexin
- Calnexin
- Antigen synonyms:
- major histocompatibility complex class I antigen-binding protein p88|CNX|FLJ26570|P90|IP90
- major histocompatibility complex class I antigen-binding protein p88|CNX|FLJ26570|P90|IP90
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Protein A affinity purified
- Protein A affinity purified
- Storage buffer:
- PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
- PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide