About this item
The 90 kDa molecular chaperone family includes 90 kDa heat shock protein Hsp90 and 94 kDa glucose-regulated protein grp94, both major molecular chaperones of the cytosol and the endoplasmic reticulum. Mammalian cells contain isoforms Hsp90 alpha and Hsp90 beta, encoded by separate genes. The amino acid sequences of human and yeast Hsp90 alpha are 85% and 90% homologous to those of Hsp90 beta, respectively. All known members of the Hsp90 protein family are highly conserved, especially in the N -terminal and C-terminal regions containing independent chaperone sites with different substrate specificity. These ubiquitous and highly conserved proteins account for 1-2% of all cellular proteins in most cells. Hsp90 functions as part of the cell
- For Western Blot
Recognizes human and chicken HSP90alpha. Detects a band of ~90kDa by Western blot.
Type: Primary
Antigen: HSP90A
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Specifications
- Catalog No:
- ENZOADISPA840HRPF
- ENZOADISPA840HRPD
- Antigen symbol:
- HSP90A
- HSP90A
- Antigen name:
- Heat Shock Protein 90 alpha
- Heat Shock Protein 90 alpha
- Antigen synonyms:
- HSP84|HSP90-BETA|heat shock 90kDa protein 1|heat shock 90kD protein|D6S182|HSPCB|HSPC2|HSPCA|heat shock protein HSP 90-beta|alpha|heat shock 90kD protein 1|heat shock 84 kDa|Hsp89|HSP86|renal carcinoma antigen NY-REN-38|HSP 86|heat shock 86 kDa|HSPCAL1|HSP 90|HSP90B|HSPN|Hsp90|beta|alpha-like 4|HSPC1|heat shock protein HSP 90-alpha|90-kda heat shock protein beta HSP90 beta|HSP90N|HSPCAL4|Hsp90aa1|LAP2|HSP89A|HSP90A|HSP 84|HSP90AB1|heat shock protein beta
- HSP84|HSP90-BETA|heat shock 90kDa protein 1|heat shock 90kD protein|D6S182|HSPCB|HSPC2|HSPCA|heat shock protein HSP 90-beta|alpha|heat shock 90kD protein 1|heat shock 84 kDa|Hsp89|HSP86|renal carcinoma antigen NY-REN-38|HSP 86|heat shock 86 kDa|HSPCAL1|HSP 90|HSP90B|HSPN|Hsp90|beta|alpha-like 4|HSPC1|heat shock protein HSP 90-alpha|90-kda heat shock protein beta HSP90 beta|HSP90N|HSPCAL4|Hsp90aa1|LAP2|HSP89A|HSP90A|HSP 84|HSP90AB1|heat shock protein beta
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Host:
- Rat
- Rat
- Isotype:
- IgG2a
- IgG2a
- Western blot:
- Yes
- Yes
- Form:
- Liquid, in PBS, pH 7,2, containing 0,09% sodium azide diluted in proprietary stabiliser solution
- Liquid, in PBS, pH 7,2, containing 0,09% sodium azide diluted in proprietary stabiliser solution
- Purification:
- Protein G affinity purified
- Protein G affinity purified