Order Entry
Austria
ContactUsLinkComponent
Anti-HSP90A Rat Monoclonal Antibody
Anti-HSP90A Rat Monoclonal Antibody
Supplier:  ENZO LIFE SCIENCES
Certificates
Anti-HSP90A Rat Monoclonal Antibody
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Catalog No:
  • ENZOADISPA840J
  • ENZOADISPA840F
  • ENZOADISPA840D
  • Antigen symbol:
  • HSP90A
  • HSP90A
  • HSP90A
  • Antigen name:
  • Heat Shock Protein 90 alpha
  • Heat Shock Protein 90 alpha
  • Heat Shock Protein 90 alpha
  • Antigen synonyms:
  • HSP84|HSP90-BETA|heat shock 90kDa protein 1|heat shock 90kD protein|D6S182|HSPCB|HSPC2|HSPCA|heat shock protein HSP 90-beta|alpha|heat shock 90kD protein 1|heat shock 84 kDa|Hsp89|HSP86|renal carcinoma antigen NY-REN-38|HSP 86|heat shock 86 kDa|HSPCAL1|HSP 90|HSP90B|HSPN|Hsp90|beta|alpha-like 4|HSPC1|heat shock protein HSP 90-alpha|90-kda heat shock protein beta HSP90 beta|HSP90N|HSPCAL4|Hsp90aa1|LAP2|HSP89A|HSP90A|HSP 84|HSP90AB1|heat shock protein beta
  • HSP84|HSP90-BETA|heat shock 90kDa protein 1|heat shock 90kD protein|D6S182|HSPCB|HSPC2|HSPCA|heat shock protein HSP 90-beta|alpha|heat shock 90kD protein 1|heat shock 84 kDa|Hsp89|HSP86|renal carcinoma antigen NY-REN-38|HSP 86|heat shock 86 kDa|HSPCAL1|HSP 90|HSP90B|HSPN|Hsp90|beta|alpha-like 4|HSPC1|heat shock protein HSP 90-alpha|90-kda heat shock protein beta HSP90 beta|HSP90N|HSPCAL4|Hsp90aa1|LAP2|HSP89A|HSP90A|HSP 84|HSP90AB1|heat shock protein beta
  • HSP84|HSP90-BETA|heat shock 90kDa protein 1|heat shock 90kD protein|D6S182|HSPCB|HSPC2|HSPCA|heat shock protein HSP 90-beta|alpha|heat shock 90kD protein 1|heat shock 84 kDa|Hsp89|HSP86|renal carcinoma antigen NY-REN-38|HSP 86|heat shock 86 kDa|HSPCAL1|HSP 90|HSP90B|HSPN|Hsp90|beta|alpha-like 4|HSPC1|heat shock protein HSP 90-alpha|90-kda heat shock protein beta HSP90 beta|HSP90N|HSPCAL4|Hsp90aa1|LAP2|HSP89A|HSP90A|HSP 84|HSP90AB1|heat shock protein beta
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rat
  • Rat
  • Rat
  • Isotype:
  • IgG2a
  • IgG2a
  • IgG2a
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Form:
  • Liquid, in PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • Liquid, in PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • Liquid, in PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • Purification:
  • Protein G affinity purified
  • Protein G affinity purified
  • Protein G affinity purified

Specifications

Frequently Bought Together