Anti-IgG1 Mouse Monoclonal Antibody (DyLight® 488)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Isotype matched controls are useful in flow cytometry to determine a gross estimate of non-specific binding that may be observed with a specific antibody of interest. In general, an isotype control should be of the same species and isotype as the antibody of interest, used at the same concentration, and conjugated to the same fluorophore. This reagent is an IgG1 isotype mouse monoclonal antibody (Clone # MOPC-21), conjugated to DyLight™-488. It is intended for use as an isotype control antibody for flow cytometry.
Type: Primary
Antigen: IgG1
Clonality: Monoclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: pan
Specifications
- Catalog No:
- ENZOADISAB600488F
- ENZOADISAB600488D
- Antigen symbol:
- IgG1
- IgG1
- Antigen name:
- Immunoglobulin G1
- Immunoglobulin G1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- DyLight® 488
- DyLight® 488
- Reactivity:
- Multiple
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Flow cytometry:
- Yes
- Yes
- Form:
- Liquid. In PBS, pH 7,2, containing 0,09% sodium azide.
- Liquid. In PBS, pH 7,2, containing 0,09% sodium azide.
- Purification:
- Protein A affinity purified
- Protein A affinity purified
Specifications
Frequently Bought Together