About this item
Heme Oxygenase-1 (HO-1) also known as Hsp32, is the inducible isoform of heme oxygenase that catalyzes the NADPH, oxygen, and cytochrome P450 reductase dependent oxidation of heme to carbon monoxide, ferrous iron and biliverdin which is rapidly reduced to bilirubin. These products of the HO reaction have important physiological effects: carbon monoxide is a potent vasodilator and has been implicated to be a physiological regulator of cGMP and vascular tone; biliverdin and its product bilirubin are potent antioxidants; "free" iron increases oxidative stress and regulates the expression of many mRNAs (e.g., DCT-1, ferritin and transferring receptor) by affecting the conformation of iron regulatory protein (IRP)-1 and its binding to iron regulatory elements (IREs) in the 5'- or 3'- UTRs of the mRNAs. To date, three identified heme oxygenase isoforms are part of the HO system that catalyze heme into biliverdin and carbon monoxide. These are inducible HO-1 or Hsp32, constitutive HO-2 that is abundant in the brain and testis, and HO-3 which is related to HO-2 but is the product of a different gene. The HO system is the rate-limiting step in heme degradation and HO activity decreases the levels of heme which is a well known potent catalyst of lipid peroxidation and oxygen radical formation.
- For Immunocytochemistry, Immunohistochemistry, Western Blot
Recognizes human, mouse, rat, bovine and dog HO-1. Detects a band of ~32kDa by Western blot.
Type: Primary
Antigen: HMOX1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- ENZOADIOSA110J
- ENZOADIOSA110F
- ENZOADIOSA110D
- Antigen symbol:
- HMOX1
- HMOX1
- HMOX1
- Antigen name:
- Heme Oxygenase 1
- Heme Oxygenase 1
- Heme Oxygenase 1
- Antigen synonyms:
- Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1
- Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1
- Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Form:
- Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
- Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
- Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
- Purification:
- Protein G affinity purified
- Protein G affinity purified
- Protein G affinity purified