Order Entry
Austria
ContactUsLinkComponent
Anti-MEK1 Rabbit Polyclonal Antibody
Anti-MEK1 Rabbit Polyclonal Antibody
Catalog # ENZOADIKAPMA010E
Supplier:  ENZO LIFE SCIENCES
Anti-MEK1 Rabbit Polyclonal Antibody
Catalog # ENZOADIKAPMA010E
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated Protein Kinase Kinase 1
  • Antigen symbol:
    MEK1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
  • Purification:
    Protein A affinity purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

MEK is a dual specificity kinase capable of phosphorylating both tyrosine and threonine residues. The MEK family, which is also known as MAP kinase kinase, phosphorylates MAP kinases on the conserved T-X-Y motif; phosphorylation of MAPK by MEK results in an increase in MAPK activity. MEK is involved in a diverse array of cellular processes such as stress-activated response, apoptosis, cytokine-induced cell proliferation, and DNA recombination during meiosis.

  • For Immunofluorescence, Immunohistochemistry, Western Blot

Recognizes human, mouse, rat and Xenopus MEK1. Detect a band of ~48kDa by Western blot.

Type: Primary
Antigen: MEK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human