Order Entry
Austria
ContactUsLinkComponent
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (V/V):
    68 --> 72 %
  • Density (20/4):
    0.869 --> 0.883
  • n 20/D:
    1.137 --> 1.375
  • Evaporation residue:
    Max. 20 ppm
  • Methanol:
    Max. 0.1 %
  • Fe (Iron):
    Max. 1 ppm
  • Pb (Lead):
    Max. 5 ppm

Specification Test Results

Frequently Bought Together