Order Entry
Austria
ContactUsLinkComponent
Human recombinant TrkA-Fc Chimera (from HEK293 cells)
Human recombinant TrkA-Fc Chimera (from HEK293 cells)
Supplier:  Biosensis
Certificates
undefined
Human recombinant TrkA-Fc Chimera (from HEK293 cells)
Supplier:  Biosensis

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilized product should be stored at 2 to 8 °C. Following reconstitution, short-term storage at 4 °C is recommended and longer-term storage of aliquots at -18 to –20 °C. Repeated freeze thawing is not recommended.
  • Gene ID:
    P04629 NTRK1_HUMAN;
  • Reconstitution instructions:
    Spin vial briefly before opening. Reconstitute in 100 uL sterile PBS, pH 7.4 to give a concentration of 0.5 mg/mL.
  • Protein synonyms:
    High affinity nerve growth factor receptor|Trk-A|Tropomyosin-related kinase receptor|TRK1 transforming tyrosine kinase protein|Neurotrophic tyrosine kinase receptor type 1|NTRK1|TRK|p140-TrkA
  • Protein/peptide name:
    TrkA-Fc Chimera
  • Purity:
    > 95% by SDS-PAGE
  • Formulation:
    Lyophilised
  • Shipping temperature:
    ambient

Specifications

Frequently Bought Together