Order Entry
Austria
ContactUsLinkComponent
Saponaria officinalis Native Saporin (from Saponaria officinalis seeds)
Saponaria officinalis Native Saporin (from Saponaria officinalis seeds)
Catalog # BSENPE-001-100
Supplier:  Biosensis
CAS Number:  
undefined
Saponaria officinalis Native Saporin (from Saponaria officinalis seeds)
Catalog # BSENPE-001-100
Supplier:  Biosensis
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Native
  • Source:
    Saponaria officinalis seeds
  • Species:
    Saponaria officinalis
  • Storage conditions:
    Keep lyophilized protein at 2-8ºC. Aliquot and keep at -20ºC for long-term storage. For short term keep aliquots at 2-8ºC. Avoid repetitive freeze/thaw cycles.
  • Gene ID:
    P20656 RIP6_SAPOF
  • Reconstitution instructions:
    Saporin may be reconstituted in the buffer of choice for example PBS. Addition of 100 uL buffer yields a saporin concentration of 1 mg/mL.
  • Protein synonyms:
    SO-6|Common soapwort|Saponaria officinalis|rRNA N-glycosidase|SAP-6|ribosome inactivating protein saporin-6
  • Protein/peptide name:
    Saporin
  • Purity:
    > 95% by SDS-PAGE
  • Molecular weight:
    29.5 kDa
  • Formulation:
    Lyophilised
  • Shipping temperature:
    ambient

Specifications

About this item

Saporin is a ribosome-inactivating protein (RIP) of type I. This monomeric RNA N-glycosidase purified from seeds of the plant Saponaria officinalis also known as Soapwort, is capable of specific depurination of eukaryotic ribosomes thus arresting protein synthesis. No ligand has been identified in saporin hence its inability to transverse the cell membrane. Due to its toxicity and stability of the structure, saporin has proven extremely useful for construction of immunotoxins.