Order Entry
Austria
ContactUsLinkComponent
BTSNM support materials, Chelex beads, 0,4 g with buffer
BTSNM support materials, Chelex beads, 0,4 g with buffer
Catalog # GENOBTNM-0024
Supplier:  G-Biosciences
CAS Number:  
BTSNM support materials, Chelex beads, 0,4 g with buffer
Catalog # GENOBTNM-0024
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Description:
    Chelex beads, 0,4 g with buffer

Specifications

About this item

Chelex beads are made of a styrene divinylbenzene copolymer containing paired iminodiacetate ions, which act as chelating groups in binding polyvalent metal ions.