Order Entry
Austria
ContactUsLinkComponent
Bicinchoninic acid disodium salt hydrate
Bicinchoninic acid disodium salt hydrate
Certificates
undefined
Bicinchoninic acid disodium salt hydrate

Specifications

  • Purity:
    ≥98% (HPLC)
  • Storage temperature:
    2...8 °C

Specifications

Frequently Bought Together