ContactUsLinkComponent
J.T.Baker®, Multipurpose Alkaline Detergent, RBS 35 concentrate
Catalog # 7348.5000
Supplier:  Avantor
J.T.Baker®, Multipurpose Alkaline Detergent, RBS 35 concentrate
Catalog # 7348.5000
Supplier:  Avantor
Specifications
  • Description:
    Multipurpose alkaline detergent, RBS 35 concentrate
  • Environmentally Preferable:
  • Pack type:
    PE bottle
  • Pk:
    5 L
  • pH range:
    11,8 (2%)
Specifications
About this item
RBS 35 concentrate is used for manual cleaning, soaking and ultrasonic baths. These surfactants are alkaline, foaming and insensitive to water hardness.
  • High wetting and emulsifying power
  • No risk of calcareous deposits when diluted in tap water
  • Total rinse
Suitable for cleaning glassware, laboratory equipments and for cleaning parts made of glass, stainless steel, injection moulds and production equipment.
Typical concentration for use: 2%