Orders LinkContactUsLinkComponent
Stacking containers, PP, LogiLine®
Stacking containers, PP, LogiLine®
Supplier:   SEMADENI
Stacking containers, PP, LogiLine®
Supplier:  SEMADENI
Certificates
Stacking containers, PP, LogiLine®
16 Options Available Below
Product Details & Documents
Made of polypropylene. These storage containers feature a solid design and an almost indestructible material.
  • Exceptionally long-lasting
  • Durable allrounder for storage and transportation
  • Temperature resistant up to +80 °C (short term up to +100 °C)
  • Suitable for quiet transport on conveyors such as roller conveyors and ball tables
The closed LogiLine container is perfectly adapted for use as a heavy duty transport container or for storing various goods in manual handling.
The LogiLine container with flat bottom is especially well suited to smooth transport on conveyor systems such as roller tracks and ball tables. Flat bottom containers have about 10 extra mm of usable height. They cannot be be stacked on containers that are the next basic size smaller.
The LogiLine container with SGL bottom meets SGL recommendations in terms of design, mechanical characteristics, and dimensional stability. Optimum usable volume, only 2 mm conicity.
Food contact approved.
Caution: Please note: The "Height" mentioned below indicates the usable height.
Product Family Options
Product Information
  • Colour
    Type
    W×D×H
    Height
    Availability
    Price
  • Grey
    With flat bottom and four handle openings
    600×400×412 mm
    393 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 123,00 €
    123,00 €
    1 * 1 ST
       123,00 €
  • Grey
    With flat bottom
    400×300×100 mm
    83 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,50 €
    32,50 €
    1 * 1 ST
       32,50 €
  • Grey
    With flat bottom
    400×300×145 mm
    128 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,40 €
    38,40 €
    1 * 1 ST
       38,40 €
  • Grey
    With flat bottom
    600×400×278 mm
    260 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,50 €
    94,50 €
    1 * 1 ST
       94,50 €
  • Grey
    With flat bottom
    400×300×212 mm
    195 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 49,70 €
    49,70 €
    1 * 1 ST
       49,70 €
  • Grey
    With flat bottom
    300×200×100 mm
    83 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,70 €
    24,70 €
    1 * 1 ST
       24,70 €
  • Grey
    With SGL bottom
    400×300×53 mm
    31 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,60 €
    29,60 €
    1 * 1 ST
       29,60 €
  • Grey
    With SGL bottom and two handle openings
    600×400×145 mm
    166 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 71,50 €
    71,50 €
    1 * 1 ST
       71,50 €
  • Grey
    With SGL bottom and two handle opening
    600×400×278 mm
    249 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,50 €
    94,50 €
    1 * 1 ST
       94,50 €
  • Grey
    With SGL bottom
    400×300×145 mm
    117 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,30 €
    37,30 €
    1 * 1 ST
       37,30 €
  • Grey
    With SGL bottom
    600×400×212 mm
    183 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,70 €
    86,70 €
    1 * 1 ST
       86,70 €
  • Grey
    With flat bottom
    300×200×145 mm
    128 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,50 €
    26,50 €
    1 * 1 ST
       26,50 €
  • Grey
    With SGL bottom
    600×400×320 mm
    291 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 97,70 €
    97,70 €
    1 * 1 ST
       97,70 €
  • Grey
    With SGL bottom
    600×400×100 mm
    72 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,20 €
    66,20 €
    1 * 1 ST
       66,20 €
  • Grey
    With flat bottom
    600×400×145 mm
    128 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 71,50 €
    71,50 €
    1 * 1 ST
       71,50 €
  • Grey
    With SGL bottom
    600×400×56 mm
    28 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,20 €
    36,20 €
    1 * 1 ST
       36,20 €
This product has been blocked by your organization. Please contact your purchasing department for more information