Order Entry
Austria
ContactUsLinkComponent
E. coli Apotryptophanase
E. coli Apotryptophanase
Catalog # SIALA6007-25MG
Supplier:  Merck
CAS Number:  
E. coli Apotryptophanase
Catalog # SIALA6007-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Species:
    E. coli
  • Form:
    White to light yellow
  • Enzyme name:
    Apotryptophanase
  • Enzyme activity:
    7000 - 15000 U/mgS
  • Enzyme synonyms:
    L-Tryptophan indole-lyase (deaminating), tryptophanase from Escherichia coli
  • Concentration:
    0,90 - 1,10 mg/ml

Specifications

About this item

Apotryptophanase isolated from Escherichia coli is used for the quantitative determination of pyridoxal-phosphate.

  • Lyophilised powder
  • Soluble

Unit definition: One unit releases 1 μg of indole from L-tryptophan in 10 min at pH 8,3 at 37 °C in the presence of 0,04 mM pyridoxal-5-phosphate.